- Directed Differentiation of Human iPSCs into Microglia-Like Cells Using Defined Transcription Factors.
Directed Differentiation of Human iPSCs into Microglia-Like Cells Using Defined Transcription Factors.
Methods in molecular biology (Clifton, N.J.) (2023-06-10)
Shih-Wei Chen, Yu-Hui Wong
PMID37300766
ABSTRACT
The generation of a homogeneous population of microglia from human induced pluripotent stem cells (hiPSCs) is crucial to modeling neurological disorders, as well as the carrying out of drug screening and toxicity testing. Here, we provide a stepwise protocol for the simple, robust, and efficient differentiation of hiPSCs into microglia-like cells (iMGs) by overexpression of SPI1 and CEBPA. This protocol details hiPSC culture, lentivirus production, lentivirus delivery, and, finally, the differentiation and validation of the iMG cells.