Merck
  • Genetics of mirror movements identifies a multifunctional complex required for Netrin-1 guidance and lateralization of motor control.
Genetics of mirror movements identifies a multifunctional complex required for Netrin-1 guidance and lateralization of motor control.
Science advances (2023-05-12)
Sabrina Schlienger, Patricia T Yam, Nursen Balekoglu, Hugo Ducuing, Jean-Francois Michaud, Shirin Makihara, Daniel K Kramer, Baoyu Chen, Alfonso Fasano, Alfredo Berardelli, Fadi F Hamdan, Guy A Rouleau, Myriam Srour, Frederic Charron
ABSTRACT
Mirror movements (MM) disorder is characterized by involuntary movements on one side of the body that mirror intentional movements on the opposite side. We performed genetic characterization of a family with autosomal dominant MM and identified ARHGEF7, a RhoGEF, as a candidate MM gene. We found that Arhgef7 and its partner Git1 bind directly to Dcc. Dcc is the receptor for Netrin-1, an axon guidance cue that attracts commissural axons to the midline, promoting the midline crossing of axon tracts. We show that Arhgef7 and Git1 are required for Netrin-1-mediated axon guidance and act as a multifunctional effector complex. Arhgef7/Git1 activates Rac1 and Cdc42 and inhibits Arf1 downstream of Netrin-1. Furthermore, Arhgef7/Git1, via Arf1, mediates the Netrin-1-induced increase in cell surface Dcc. Mice heterozygous for Arhgef7 have defects in commissural axon trajectories and increased symmetrical paw placements during skilled walking, a MM-like phenotype. Thus, we have delineated how ARHGEF7 mutation causes MM.
Sigma-Aldrich
Anti-Beta Pix, SH3 Domain Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Rac1 Antibody, clone 23A8, clone 23A8, Upstate®, from mouse