Merck
  • Human early syncytiotrophoblasts are highly susceptible to SARS-CoV-2 infection.
Human early syncytiotrophoblasts are highly susceptible to SARS-CoV-2 infection.
Cell reports. Medicine (2022-12-11)
Degong Ruan, Zi-Wei Ye, Shuofeng Yuan, Zhuoxuan Li, Weiyu Zhang, Chon Phin Ong, Kaiming Tang, Timothy Theodore Ka Ki Tam, Jilong Guo, Yiyi Xuan, Yunying Huang, Qingqing Zhang, Cheuk-Lun Lee, Liming Lu, Philip C N Chiu, William S B Yeung, Fang Liu, Dong-Yan Jin, Pentao Liu
ABSTRACT
Direct in vivo investigation of human placenta trophoblast's susceptibility to SARS-CoV-2 is challenging. Here we report that human trophoblast stem cells (hTSCs) and their derivatives are susceptible to SARS-CoV-2 infection, which reveals heterogeneity in hTSC cultures. Early syncytiotrophoblasts (eSTBs) generated from hTSCs have enriched transcriptomic features of peri-implantation trophoblasts, express high levels of angiotensin-converting enzyme 2 (ACE2), and are productively infected by SARS-CoV-2 and its Delta and Omicron variants to produce virions. Antiviral drugs suppress SARS-CoV-2 replication in eSTBs and antagonize the virus-induced blockage of STB maturation. Although less susceptible to SARS-CoV-2 infection, trophoblast organoids originating from hTSCs show detectable viral replication reminiscent of the uncommon placental infection. These findings implicate possible risk of COVID-19 infection in peri-implantation embryos, which may go unnoticed. Stem cell-derived human trophoblasts such as eSTBs can potentially provide unlimited amounts of normal and genome-edited cells and facilitate coronavirus research and antiviral discovery.
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Bovine Serum Albumin solution, 35% in DPBS, sterile-filtered, BioXtra, suitable for cell culture