Merck
  • DRP1 levels determine the apoptotic threshold during embryonic differentiation through a mitophagy-dependent mechanism.
DRP1 levels determine the apoptotic threshold during embryonic differentiation through a mitophagy-dependent mechanism.
Developmental cell (2022-05-22)
Barbara Pernaute, Salvador Pérez-Montero, Juan Miguel Sánchez Nieto, Aida Di Gregorio, Ana Lima, Katerina Lawlor, Sarah Bowling, Gianmaria Liccardi, Alejandra Tomás, Pascal Meier, Hiromi Sesaki, Guy A Rutter, Ivana Barbaric, Tristan A Rodríguez
ABSTRACT
The changes that drive differentiation facilitate the emergence of abnormal cells that need to be removed before they contribute to further development or the germline. Consequently, in mice in the lead-up to gastrulation, ∼35% of embryonic cells are eliminated. This elimination is caused by hypersensitivity to apoptosis, but how it is regulated is poorly understood. Here, we show that upon exit of naive pluripotency, mouse embryonic stem cells lower their mitochondrial apoptotic threshold, and this increases their sensitivity to cell death. We demonstrate that this enhanced apoptotic response is induced by a decrease in mitochondrial fission due to a reduction in the activity of dynamin-related protein 1 (DRP1). Furthermore, we show that in naive pluripotent cells, DRP1 prevents apoptosis by promoting mitophagy. In contrast, during differentiation, reduced mitophagy levels facilitate apoptosis. Together, these results indicate that during early mammalian development, DRP1 regulation of mitophagy determines the apoptotic response.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Bak (NT) Antibody, from rabbit, purified by affinity chromatography