Merck
  • Selective binding of retrotransposons by ZFP352 facilitates the timely dissolution of totipotency network.
Selective binding of retrotransposons by ZFP352 facilitates the timely dissolution of totipotency network.
Nature communications (2023-06-21)
Zhengyi Li, Haiyan Xu, Jiaqun Li, Xiao Xu, Junjiao Wang, Danya Wu, Jiateng Zhang, Juan Liu, Ziwei Xue, Guankai Zhan, Bobby Cheng Peow Tan, Di Chen, Yun-Shen Chan, Huck Hui Ng, Wanlu Liu, Chih-Hung Hsu, Dan Zhang, Yang Shen, Hongqing Liang
ABSTRACT
Acquisition of new stem cell fates relies on the dissolution of the prior regulatory network sustaining the existing cell fates. Currently, extensive insights have been revealed for the totipotency regulatory network around the zygotic genome activation (ZGA) period. However, how the dissolution of the totipotency network is triggered to ensure the timely embryonic development following ZGA is largely unknown. In this study, we identify the unexpected role of a highly expressed 2-cell (2C) embryo specific transcription factor, ZFP352, in facilitating the dissolution of the totipotency network. We find that ZFP352 has selective binding towards two different retrotransposon sub-families. ZFP352 coordinates with DUX to bind the 2C specific MT2_Mm sub-family. On the other hand, without DUX, ZFP352 switches affinity to bind extensively onto SINE_B1/Alu sub-family. This leads to the activation of later developmental programs like ubiquitination pathways, to facilitate the dissolution of the 2C state. Correspondingly, depleting ZFP352 in mouse embryos delays the 2C to morula transition process. Thus, through a shift of binding from MT2_Mm to SINE_B1/Alu, ZFP352 can trigger spontaneous dissolution of the totipotency network. Our study highlights the importance of different retrotransposons sub-families in facilitating the timely and programmed cell fates transition during early embryogenesis.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Prosurfactant Protein B Antibody, reacts with both CT and NT, serum, Chemicon®