Merck
  • Increased Circulating ADMA in Young Male Rats Caused Cognitive Deficits and Increased Intestinal and Hippocampal NLRP3 Inflammasome Expression and Microbiota Composition Alterations: Effects of Resveratrol.
Increased Circulating ADMA in Young Male Rats Caused Cognitive Deficits and Increased Intestinal and Hippocampal NLRP3 Inflammasome Expression and Microbiota Composition Alterations: Effects of Resveratrol.
Pharmaceuticals (Basel, Switzerland) (2023-06-28)
Mei-Hsin Hsu, Yi-Chuan Huang, Yu-Chieh Chen, Jiunn-Ming Sheen, Li-Tung Huang
ABSTRACT
Endothelial dysfunction is characterized by disturbances in nitric oxide (NO) bioavailability and increased circulating asymmetric dimethylarginine (ADMA) due to the enormous release of free radicals. Increased circulating ADMA may cause endothelial dysfunction and a variety of clinical disorders, such as liver and kidney disease. Young male Sprague-Dawley rats at postnatal day 17 ± 1 received continuous ADMA infusion via an intraperitoneal pump to induce endothelial dysfunction. Four groups of rats (n = 10 per group) were allocated: control, control and resveratrol, ADMA infusion, and ADMA infusion and resveratrol groups. Spatial memory, NLR family pyrin-domain-containing 3 (NLRP3) inflammasome, cytokine expression, tight junction proteins in the ileum and dorsal hippocampus, and microbiota composition were examined. We found cognitive deficits; increased NLRP3 inflammasome in the plasma, ileum, and dorsal hippocampus; decreased ileum and dorsal hippocampal cytokine activation and tight junction proteins; and microbiota composition alterations in the ADMA-infusion young male rats. Resveratrol had beneficial effects in this context. In conclusion, we observed NLRP3 inflammasome activation in peripheral and central dysbiosis in young male rats with increased circulating ADMA, and found that resveratrol had beneficial effects. Our work adds to the mounting evidence that inhibiting systemic inflammation is a promising therapeutic avenue for cognition impairment, probably via the gut-brain axis.