Merck
  • Identification of environmental factors that promote intestinal inflammation.
Identification of environmental factors that promote intestinal inflammation.
Nature (2022-10-21)
Liliana M Sanmarco, Chun-Cheih Chao, Yu-Chao Wang, Jessica E Kenison, Zhaorong Li, Joseph M Rone, Claudia M Rejano-Gordillo, Carolina M Polonio, Cristina Gutierrez-Vazquez, Gavin Piester, Agustin Plasencia, Lucinda Li, Federico Giovannoni, Hong-Gyun Lee, Camilo Faust Akl, Michael A Wheeler, Ivan Mascanfroni, Merja Jaronen, Moneera Alsuwailm, Patrick Hewson, Ada Yeste, Brian M Andersen, Diana G Franks, Chien-Jung Huang, Millicent Ekwudo, Emily C Tjon, Veit Rothhammer, Maisa Takenaka, Kalil Alves de Lima, Mathias Linnerbauer, Lydia Guo, Ruxandra Covacu, Hugo Queva, Pedro Henrique Fonseca-Castro, Maha Al Bladi, Laura M Cox, Kevin J Hodgetts, Mark E Hahn, Alexander Mildner, Joshua Korzenik, Russ Hauser, Scott B Snapper, Francisco J Quintana
ABSTRACT
Genome-wide association studies have identified risk loci linked to inflammatory bowel disease (IBD)1-a complex chronic inflammatory disorder of the gastrointestinal tract. The increasing prevalence of IBD in industrialized countries and the augmented disease risk observed in migrants who move into areas of higher disease prevalence suggest that environmental factors are also important determinants of IBD susceptibility and severity2. However, the identification of environmental factors relevant to IBD and the mechanisms by which they influence disease has been hampered by the lack of platforms for their systematic investigation. Here we describe an integrated systems approach, combining publicly available databases, zebrafish chemical screens, machine learning and mouse preclinical models to identify environmental factors that control intestinal inflammation. This approach established that the herbicide propyzamide increases inflammation in the small and large intestine. Moreover, we show that an AHR-NF-κB-C/EBPβ signalling axis operates in T cells and dendritic cells to promote intestinal inflammation, and is targeted by propyzamide. In conclusion, we developed a pipeline for the identification of environmental factors and mechanisms of pathogenesis in IBD and, potentially, other inflammatory diseases.
Sigma-Aldrich
S-Methyl-L-thiocitrulline acetate salt, ≥98% (TLC)
Sigma-Aldrich
Bay 11-7082, ≥98% (HPLC), powder
Roche
DNase I, grade II, from bovine pancreas
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution