Merck
  • Identification and characterization of circulating immune complexes in IgA nephropathy.
Identification and characterization of circulating immune complexes in IgA nephropathy.
Science advances (2022-10-29)
Yasuyuki Matsumoto, Rajindra P Aryal, Jamie Heimburg-Molinaro, Simon S Park, Walter J Wever, Sylvain Lehoux, Kathrin Stavenhagen, Joanna A E van Wijk, Irma Van Die, Arlene B Chapman, Elliot L Chaikof, Richard D Cummings
ABSTRACT
The underlying pathology of immunoglobulin A (IgA) nephropathy (IgAN), the most common glomerulonephritis worldwide, is driven by the deposition of immune complexes containing galactose-deficient IgA1 [Tn(+)IgA1] in the glomerular mesangium. Here, we report that novel anti-Tn circulating immune complexes (anti-Tn CICs) contain predominantly IgM, representing large macromolecular complexes of ~1.2 megadaltons to several megadalton sizes together with Tn(+)IgA1 and some IgG. These complexes are significantly elevated in sera of patients with IgAN, which contains higher levels of complement C3, compared to healthy individuals. Anti-Tn CICs are bioactive and induce specific proliferation of human renal mesangial cells. We found that these anti-Tn CICs can be dissociated with small glycomimetic compounds, which mimic the Tn antigen of Tn(+)IgA1, releasing IgA1 from anti-Tn CICs. This glycomimetic compound can also significantly inhibit the proliferative activity of anti-Tn CICs of patients with IgAN. These findings could enhance both the diagnosis of IgAN and its treatment, as specific drug treatments are now unavailable.
Sigma-Aldrich
Lectin from Helix pomatia, lyophilized powder, salt, free
Sigma-Aldrich
Lectin from Helix pomatia, biotin conjugate, lyophilized powder
Roche
Neuraminidase (Sialidase), from Arthrobacter ureafaciens