Merck
  • Effects of Caloric Restriction on Spatial Object Recognition Memory, Hippocampal Neuron Loss and Neuroinflammation in Aged Rats.
Effects of Caloric Restriction on Spatial Object Recognition Memory, Hippocampal Neuron Loss and Neuroinflammation in Aged Rats.
Nutrients (2023-04-14)
Marta Portero-Tresserra, Neus Galofré-López, Elisabet Pallares, Claudia Gimenez-Montes, Carlos Barcia, Roser Granero, Divka Rojic-Becker, Anna Vale-Martínez, Margarita Martí-Nicolovius, Gemma Guillazo-Blanch
ABSTRACT
Age-related neurobiological changes significantly affect hippocampal structure and function, such that the main cognitive impairments associated with aging are related to the integrity of this brain structure, including the deterioration in spatial object recognition (SOR) memory. Previous studies have shown that intrinsic factors such as neuroinflammation, as well as lifestyle factors such as diet, can affect aging-associated brain functions and cognitive performance. In this regard, caloric restriction (CR) produces beneficial effects on health and life expectancy, although its ability to slow down age-dependent effects on cognitive decline and hippocampus (HPC) functioning remains unclear. Therefore, we set out to evaluate the effects of CR on SOR memory in aged male Wistar rats, as well as those on hippocampal neuron loss, neurogenesis and inflammation. The data show that CR in aged rats attenuates the decline in SOR memory, age-associated hippocampal neuron loss, and age-dependent microglial activation. Furthermore, we found a significant reduction in neurogenesis in the dentate gyrus of the old animals relative to adult rats. These findings support the positive effect of CR on SOR memory, suggesting that it dampens hippocampal neuronal loss and reduces proinflammatory activity.