Merck
Chaperone-mediated autophagy regulates adipocyte differentiation.
Science advances (2022-11-17)
Susmita Kaushik, Yves R Juste, Kristen Lindenau, Shuxian Dong, Adrián Macho-González, Olaya Santiago-Fernández, Mericka McCabe, Rajat Singh, Evripidis Gavathiotis, Ana Maria Cuervo
ABSTRACT
Adipogenesis is a tightly orchestrated multistep process wherein preadipocytes differentiate into adipocytes. The most studied aspect of adipogenesis is its transcriptional regulation through timely expression and silencing of a vast number of genes. However, whether turnover of key regulatory proteins per se controls adipogenesis remains largely understudied. Chaperone-mediated autophagy (CMA) is a selective form of lysosomal protein degradation that, in response to diverse cues, remodels the proteome for regulatory purposes. We report here the activation of CMA during adipocyte differentiation and show that CMA regulates adipogenesis at different steps through timely degradation of key regulatory signaling proteins and transcription factors that dictate proliferation, energetic adaptation, and signaling changes required for adipogenesis.
Sigma-Aldrich
Anti-DLK1 Antibody, clone 3A10, ascites fluid, clone 3A10, from mouse
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%