Merck
  • Characterisation of IL-23 receptor antagonists and disease relevant mutants using fluorescent probes.
Characterisation of IL-23 receptor antagonists and disease relevant mutants using fluorescent probes.
Nature communications (2023-05-20)
Charles S Lay, Albert Isidro-Llobet, Laura E Kilpatrick, Peter D Craggs, Stephen J Hill
ABSTRACT
Association of single nucleotide polymorphisms in the IL-23 receptor with several auto-inflammatory diseases, led to the heterodimeric receptor and its cytokine-ligand IL-23, becoming important drug targets. Successful antibody-based therapies directed against the cytokine have been licenced and a class of small peptide antagonists of the receptor have entered clinical trials. These peptide antagonists may offer therapeutic advantages over existing anti-IL-23 therapies, but little is known about their molecular pharmacology. In this study, we use a fluorescent version of IL-23 to characterise antagonists of the full-length receptor expressed by living cells using a NanoBRET competition assay. We then develop a cyclic peptide fluorescent probe, specific to the IL23p19:IL23R interface and use this molecule to characterise further receptor antagonists. Finally, we use the assays to study the immunocompromising C115Y IL23R mutation, demonstrating that the mechanism of action is a disruption of the binding epitope for IL23p19.