Merck
Issues with RNF43 antibodies to reliably detect intracellular location.
PloS one (2023-04-07)
Shanshan Li, Ruyi Zhang, Marla Lavrijsen, Thierry P P van den Bosch, Maikel P Peppelenbosch, Ron Smits
ABSTRACT
RNF43 is an important negative regulator of β-catenin signaling by removing Wnt-receptors from the membrane. It is often mutated in cancers, leading to aberrant Wnt-dependent nuclear translocation of β-catenin. RNF43 has also been suggested to regulate β-catenin signaling directly within the nucleus, among other proposed nuclear functions. Given the importance of RNF43 in regulating Wnt/β-catenin signaling and its potential therapeutic relevance, a proper understanding of RNF43 biology is required. However, the presumed nuclear location is mainly based on available antibodies. These same antibodies have also been used extensively for immunoblotting or immunohistochemical purposes. However, a proper evaluation of their quality to reliably detect endogenous RNF43 has not been performed. Here, using genome editing we have generated a cell line that entirely misses RNF43 exons 8 and 9, encoding the epitopes of commonly used RNF43 antibodies. Using this clone in addition to various other cell line tools, we show that four RNF43 antibodies only yield non-specific signals when applied in immunoblotting, immunofluorescence and immunohistochemical experiments. In other words, they cannot reliably detect endogenous RNF43. Our results suggest that the nuclear staining patterns are an antibody artifact and that RNF43 is unlikely to localize within the nucleus. More generally, reports using RNF43 antibodies should be interpreted with caution, at least for the RNF43 protein aspects described in these papers.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)