Merck
  • IFN-γ differential expression in the hypothalamus-pituitary-ovary axis of thyroidectomized rats.
IFN-γ differential expression in the hypothalamus-pituitary-ovary axis of thyroidectomized rats.
BMC endocrine disorders (2022-12-14)
Jingjie Wei, Yan Liang, Ningbo Jiang, Ge Hu
ABSTRACT
IFN-γ is critical for both thyroid and ovarian function, while thyroxine, secreted from the thyroid gland, regulates the ovarian function via the hypothalamus-pituitary -ovary axis. However, the effect of thyroxine on INF-γ involved in the regulation of hypothalamic pituitary ovarian axis ovarian function is hitherto unknown. Therefore, we set up three groups including a sham-operated group, an experimental thyroidectomized group, and an experimental thyroidectomized group treated with T4 to reveal the IFN-γ expression levels in the in the hypothalamus, pituitary gland, and ovary by immunohistochemical staining, RT-PCR, and Western blotting. IFN-γ-like immunoreactive-positive substances were visualized in the hypothalamus, pituitary gland, and ovary, which were located mainly in the cytoplasm of the hypothalamic neurons anterior pituitary cells, luteal cells, and theca cells in the ovary of hypothyroidism rats, respectively. RT-PCR and Western blotting showed that the rats in the experimental thyroidectomized group treated with T4 had significantly elevated expression of IFN-γ at both the mRNA and protein levels. Thyroxine affects the expression of IFN-γ in the thalamus-pituitary-ovarian axis, which may influence the secretion of IFN-γ to regulate ovarian function during hypothyroidism. This work highlights the potential effect of thyroxine on the involvement of INF-γ in the modulation of the ovarian function in the hypothalamic-pituitary-ovarian axis.