Merck
  • Pathological 25 kDa C-Terminal Fragments of TDP-43 Are Present in Lymphoblastoid Cell Lines and Extracellular Vesicles from Patients Affected by Frontotemporal Lobar Degeneration and Neuronal Ceroidolipofuscinosis Carrying a GRN Mutation.
Pathological 25 kDa C-Terminal Fragments of TDP-43 Are Present in Lymphoblastoid Cell Lines and Extracellular Vesicles from Patients Affected by Frontotemporal Lobar Degeneration and Neuronal Ceroidolipofuscinosis Carrying a GRN Mutation.
International journal of molecular sciences (2022-11-27)
Sara Cimini, Sonia Bellini, Claudia Saraceno, Luisa Benussi, Roberta Ghidoni, Silvia Clara Giliani, Gianfranco Puoti, Laura Canafoglia, Giorgio Giaccone, Giacomina Rossi
ABSTRACT
Frontotemporal lobar degeneration (FTLD) is a complex disease, characterized by progressive degeneration of frontal and temporal lobes. Mutations in progranulin (GRN) gene have been found in up to 50% of patients with familial FTLD. Abnormal deposits of post-translationally-modified TAR DNA-binding protein of 43 kDa (TDP-43) represent one of the main hallmarks of the brain pathology. To investigate in peripheral cells the presence of the different TDP-43 forms, especially the toxic 25 kDa fragments, we analyzed lymphoblastoid cell lines (LCLs) and the derived extracellular vesicles (EVs) from patients carrying a GRN mutation, together with wild-type (WT) healthy controls. After characterizing EV sizes and concentrations by nanoparticle tracking analysis, we investigated the levels of different forms of the TDP-43 protein in LCLs and respective EVs by Western blot. Our results showed a trend of concentration decreasing in EVs derived from GRN-mutated LCLs, although not reaching statistical significance. A general increase in p-TDP-43 levels in GRN-mutated LCLs and EVs was observed. In particular, the toxic 25 kDa fragments of p-TDP-43 were only present in GRN-mutated LCLs and were absent in the WT controls. Furthermore, these fragments appeared to be more concentrated in EVs than in LCLs, suggesting a relevant role of EVs in spreading pathological molecules between cells.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse