Merck
  • Glutamate released by Cajal-Retzius cells impacts specific hippocampal circuits and behaviors.
Glutamate released by Cajal-Retzius cells impacts specific hippocampal circuits and behaviors.
Cell reports (2022-05-19)
Max Anstötz, Sun Kyong Lee, Gianmaria Maccaferri
ABSTRACT
The impact of Cajal-Retzius cells on the regulation of hippocampal circuits and related behaviors is unresolved. Here, we directly address this issue by impairing the glutamatergic output of Cajal-Retzius cells with the conditional ablation of vGluT2, which is their main vesicular glutamate transporter. Although two distinct conditional knockout lines do not reveal major alterations in hippocampal-layer organization and dendritic length of principal neurons or GABAergic cells, we find parallel deficits in specific hippocampal-dependent behaviors and in their putative underlying microcircuits. First, conditional knockout animals show increased innate anxiety and decreased feedforward GABAergic inhibition on dentate gyrus granule cells. Second, we observe impaired spatial memory processing, which is associated with decreased spine density and reduced AMPA/NMDA ratio of postsynaptic responses at the perforant- and entorhino-hippocampal pathways. We conclude that glutamate synaptically released by Cajal-Retzius cells is critical for the regulation of hippocampal microcircuits and specific types of behaviors.
Sigma-Aldrich
Anti-GAD 65/67, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, clone G10, Chemicon®, from mouse