Merck
  • Ubiquitin E3 ligase Atrogin-1 protein is regulated via the rapamycin-sensitive mTOR-S6K1 signaling pathway in C2C12 muscle cells.
Ubiquitin E3 ligase Atrogin-1 protein is regulated via the rapamycin-sensitive mTOR-S6K1 signaling pathway in C2C12 muscle cells.
American journal of physiology. Cell physiology (2022-06-16)
Yusuke Nishimura, Jitpisute Chunthorng-Orn, Samuel Lord, Ibrahim Musa, Peter Dawson, Lars Holm, Yu-Chiang Lai
ABSTRACT
Atrogin-1 and Muscle-specific RING finger protein 1 (MuRF1) are highly expressed in multiple conditions of skeletal muscle atrophy. The phosphoinositide 3-kinase (PI3K)/Akt/forkhead box (FoxO) signaling pathway is well known to regulate Atrogin-1 and MuRF1 gene expressions. However, Akt activation also activates the mechanistic target of rapamycin complex 1 (mTORC1), which induces skeletal muscle hypertrophy. Whether mTORC1-dependent signaling has a role in regulating Atrogin-1 and/or MuRF1 gene and protein expression is currently unclear. In this study, we showed that activation of insulin-mediated Akt signaling suppresses both Atrogin-1 and MuRF1 protein contents and that inhibition of Akt increases both Atrogin-1 and MuRF1 protein contents in C2C12 myotubes. Interestingly, inhibition of mTORC1 with a specific mTORC1 inhibitor, rapamycin, increased Atrogin-1, but not MuRF1, protein content. Furthermore, activation of AMP-activated protein kinase (AMPK), a negative regulator of the mTORC1 signaling pathway, also showed distinct time-dependent changes between Atrogin-1 and MuRF1 protein contents, suggesting differential regulatory mechanisms between Atrogin-1 and MuRF1 protein content. To further explore the downstream of mTORC1 signaling, we employed a specific S6K1 inhibitor, PF-4708671. We found that Atrogin-1 protein content was dose-dependently increased with PF-4708671 treatment, whereas MuRF1 protein content was decreased at 50 μM of PF-4708671 treatment. However, MuRF1 protein content was unexpectedly increased by PF-4708671 treatment for a longer period. Overall, our results indicate that Atrogin-1 and MuRF1 protein contents are regulated by different mechanisms, the downstream of Akt, and that Atrogin-1 protein content can be regulated by the rapamycin-sensitive mTOR-S6K1-dependent signaling pathway.
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus