Merck
  • Actin-driven chromosome clustering facilitates fast and complete chromosome capture in mammalian oocytes.
Actin-driven chromosome clustering facilitates fast and complete chromosome capture in mammalian oocytes.
Nature cell biology (2023-02-03)
Katarina Harasimov, Julia Uraji, Eike Urs Mönnich, Zuzana Holubcová, Kay Elder, Martyn Blayney, Melina Schuh
ABSTRACT
Accurate chromosome segregation during meiosis is crucial for reproduction. Human and porcine oocytes transiently cluster their chromosomes before the onset of spindle assembly and subsequent chromosome segregation. The mechanism and function of chromosome clustering are unknown. Here we show that chromosome clustering is required to prevent chromosome losses in the long gap phase between nuclear envelope breakdown and the onset of spindle assembly, and to promote the rapid capture of all chromosomes by the acentrosomal spindle. The initial phase of chromosome clustering is driven by a dynamic network of Formin-2- and Spire-nucleated actin cables. The actin cables form in the disassembling nucleus and migrate towards the nuclear centre, moving the chromosomes centripetally by interacting with their arms and kinetochores as they migrate. A cage of stable microtubule loops drives the late stages of chromosome clustering. Together, our data establish a crucial role for chromosome clustering in accurate progression through meiosis.
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder