Merck
  • Myelin-associated glycoprotein alters the neuronal secretome and stimulates the release of TGFβ and proteins that affect neural plasticity.
Myelin-associated glycoprotein alters the neuronal secretome and stimulates the release of TGFβ and proteins that affect neural plasticity.
FEBS letters (2022-09-15)
Vanessa D Stewart, Justine Cadieux, Matsya R Thulasiram, Tinsley Claire Douglas, Dennis A Drewnik, Suhaila Selamat, Ying Lao, Victor Spicer, Sari S Hannila
ABSTRACT
Myelin-associated glycoprotein (MAG) and Nogo inhibit neurite outgrowth by binding to receptors such as NgR1, PirB and LRP1, and they have also been shown to induce phosphorylation of Smad2, a key intermediate in the transforming growth factor β (TGFβ) signalling pathway. In this study, we determined that MAG and Nogo do not transactivate the TGFβ receptor through their canonical receptors or discoidin domain receptor 1, which we identified as a novel receptor for MAG and Nogo. Instead, MAG and Nogo promoted Smad2 phosphorylation by stimulating secretion of TGFβ. Proteomic analysis of the neuronal secretome revealed that MAG also regulated the secretion of proteins that affect central nervous system plasticity-inducing the secretion of S100A6, septin-7 and neurofascin 186, while inhibiting the secretion of frataxin, MAP6, syntenin-1 and GAP-43. This represents a novel function for MAG that has broad implications for the treatment for spinal cord injury.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
SB-505124 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Anti-Myelin Associated Glycoprotein Antibody, clone 513, clone 513, Chemicon®, from mouse
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, aseptically filled
Sigma-Aldrich
MBP, recombinant