Merck
  • Identification and molecular analysis of RNF31 Q622H germline polymorphism.
Identification and molecular analysis of RNF31 Q622H germline polymorphism.
Oncology letters (2022-10-25)
Seshiru Nakazawa, Ryo Mamiya, Reika Kawabata-Iwakawa, Daisuke Oikawa, Kyoichi Kaira, Fuminori Tokunaga, Sumihito Nobusawa, Yusuke Sato, Atsushi Sasaki, Toshiki Yajima, Ken Shirabe
ABSTRACT
The linear ubiquitin chain assembly complex (LUBAC), which is composed of RING finger protein 31 (RNF31), RANBP2-type and C3HC4-type zinc finger containing 1 and SHANK-associated RH domain interactor subunits, is the only ubiquitin ligase to generate Met1-linked linear ubiquitin chains. Linear ubiquitin chains regulate canonical NF-κB activation and cell death. Single nucleotide polymorphisms in RNF31, such as Q584H and Q622L, are known to cause the activated B cell-like subtype of diffuse large B cell lymphoma (ABC-DLBCL) because of enhanced LUBAC-mediated NF-κB activation. The present study identified a novel Q622H polymorphism of RNF31 in two patients with lung cancer, one of whom had concurrent ABC-DLBCL. Immunohistochemical analyses revealed that although the expression of RNF31 was elevated in both patients, only the ABC-DLBCL specimen showed increased NF-κB activation. Cancer panel analysis showed that the Q622H-related ABC-DLBCL did not harbor co-mutations that were previously reported in Q584H-/Q622L-related ABC-DLBCL. Furthermore, in contrast to Q584H and Q622L, Q622H showed no enhancement effects on LUBAC and NF-κB activity in vitro compared with wild-type RNF31. The present study's structural prediction suggested that the electrostatic interaction related to the Q622 residue may not have had an important role in LUBAC formation. In conclusion, the molecular mechanism and mutational background of RNF31 Q622H differed from that of RNF31 Q584H or Q622L. Furthermore, RNF31 Q622H appeared not to induce NF-κB activation in lung cancer.
Sigma-Aldrich
Anti-Linear Ubiquitin Antibody, clone LUB9, clone LUB9, from mouse