Merck
  • Electroacupuncture relieves neuropathic pain by inhibiting degradation of the ecto-nucleotidase PAP in the dorsal root ganglions of CCI mice.
Electroacupuncture relieves neuropathic pain by inhibiting degradation of the ecto-nucleotidase PAP in the dorsal root ganglions of CCI mice.
European journal of pain (London, England) (2022-02-10)
Xiaoxin Zhou, Wanbing Dai, Yi Qin, Siyi Qi, Yizhe Zhang, Weitian Tian, Xiyao Gu, Beijie Zheng, Jie Xiao, Weifeng Yu, Xuemei Chen, Diansan Su
ABSTRACT
Although electroacupuncture is widely used in chronic pain management, it is quite controversial due to its unclear mechanism. We hypothesised that EA alleviates pain by inhibiting degradation of the ecto-nucleotidase prostatic acid phosphatase (PAP) and facilitating ATP dephosphorylation in dorsal root ganglions (DRGs). We applied EA in male C57 mice subjected to chronic constriction injury (CCI) and assessed extracellular ATP and 5'-nucleotidease expression in DRGs. Specifically, we used a luminescence assay, quantitative reverse transcriptase-polymerase chain reaction, Western blotting, immunohistochemistry and nociceptive-related behavioural changes to gather data, and we tested for effects after PAP expression was inhibited with an adeno-associated virus (AAV). Moreover, membrane PAP degradation was investigated in cultured DRG neurons and the inhibitory effects of EA on this degradation were assessed using immunoprecipitation. EA treatment alleviated CCI surgery-induced mechanical pain hypersensitivity. Furthermore, extracellular ATP decreased significantly in both the DRGs and dorsal horn of EA-treated mice. PAP protein but not mRNA increased in L4-L5 DRGs, and inhibition of PAP expression via AAV microinjection reversed the analgesic effect of EA. Membrane PAP degradation occurred through a clathrin-mediated endocytosis pathway in cultured DRG neurons; EA treatment inhibited the phosphorylation of adaptor protein complex 2, which subsequently reduced the endocytosis of membrane PAP. EA treatment alleviated peripheral nerve injury-induced mechanical pain hypersensitivity in mice by inhibiting membrane PAP degradation via reduced endocytosis and subsequently promote ATP dephosphorylation in DRGs. In a mouse model of chronic pain, electroacupuncture treatment increased levels of prostatic acid phosphatase (PAP: an ecto-nucleotidase known to relieve pain hypersensitivity) by inhibiting PAP degradation in dorsal root ganglions. This promoted extracellular ATP dephosphorylation, inhibited glia activation and eventually alleviated peripheral nerve injury-induced mechanical pain hypersensitivity in mice. Our findings represent an important step forward in clarifying the mechanisms of pain relief afforded by acupuncture treatment.
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture