Merck
  • Dihydroartemisinin alleviates hepatic fibrosis through inducing ferroptosis in hepatic stellate cells.
Dihydroartemisinin alleviates hepatic fibrosis through inducing ferroptosis in hepatic stellate cells.
BioFactors (Oxford, England) (2021-06-16)
Zili Zhang, Xian Wang, Zilong Wang, Zhiyue Zhang, Yashi Cao, Zonghui Wei, Jiangjuan Shao, Anping Chen, Feng Zhang, Shizhong Zheng
ABSTRACT
Targeting the elimination of activated hepatic stellate cells (HSCs) and blocking excessive deposition of extracellular matrix are recognized as an effective strategy for the treatment of hepatic fibrosis. As a newly discovered programmed cell death mode, the regulatory mechanism of ferroptosis in the clearance of activated HSCs has not been fully elucidated. In the present study, we reported that the induction of ferroptosis in activated HSCs was required for dihydroartemisinin (DHA) to alleviate hepatic fibrosis. Treatment with DHA could improve the damage of hepatic fibrosis in vivo and inhibit the activation of HSCs in vitro. Interestingly, DHA treatment could trigger ferroptosis to eliminate activated HSCs characterized by iron overload, lipid ROS accumulation, glutathione depletion, and lipid peroxidation. Specific ferroptosis inhibitors ferrostatin-1 and liproxstatin-1 could impair DHA-induced ferroptosis and also damage DHA-mediated the inhibition of activated HSCs. Importantly, autophagy activation may be closely related to DHA-induced ferroptosis. ATG5 siRNA could prevent DHA-mediated autophagy activation and ferroptosis induction, whereas ATG5 plasmid could promote the effect of DHA on autophagy and ferroptosis. Of note, the upregulation of nuclear receptor coactivator 4 (NCOA4) may play a critical role in the molecular mechanism. NCOA4 siRNA could impair DHA-induced ferroptosis, whereas NCOA4 plasmid could enhance the promoting effect of DHA on ferroptosis. Overall, our study revealed the potential mechanism of DHA against hepatic fibrosis and showed that ferroptosis could be a new way to eliminate activated HSCs.
Sigma-Aldrich
Universal Antibody Dilution Buffer, Ready-To-Use, with BSA
Sigma-Aldrich
Monoclonal Anti-Rabbit Immunoglobulins–FITC antibody produced in mouse, clone RG-16, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Sigma-Aldrich
Erastin, ≥98% (HPLC)
Sigma-Aldrich
PDGF-BB, recombinant human, The Platelet Derived Growth Factor (PDGF) is a potent mitogen for a wide range of cell types including fibroblasts, smooth muscle & connective tissue.
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)