Merck
  • Chromatin Immunoprecipitation-Polymerase Chain Reaction (ChIP-PCR) Detects Methylation, Acetylation, and Ubiquitylation in S. pombe.
Chromatin Immunoprecipitation-Polymerase Chain Reaction (ChIP-PCR) Detects Methylation, Acetylation, and Ubiquitylation in S. pombe.
Methods in molecular biology (Clifton, N.J.) (2018-02-10)
Valerie Migeot, Damien Hermand
ABSTRACT
The distribution of modified histones within the fission yeast Schizosaccharomyces pombe genome is ultimately dependent upon the transcriptional activity and in turn influences the ability of the polymerases to bind and progress through the chromatin template. The Chromatin Immunoprecipitation-Polymerase Chain Reaction (ChIP-PCR) method currently provides the highest resolution, accuracy, and reproducibility to characterize histones modifications within a defined region of the genome. The following protocol details the method applied to S. pombe.
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys14) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys36) Antibody, serum, Upstate®