Merck
  • CXCL12/CXCR4 promotes proliferation, migration, and invasion of adamantinomatous craniopharyngiomas via PI3K/AKT signal pathway.
CXCL12/CXCR4 promotes proliferation, migration, and invasion of adamantinomatous craniopharyngiomas via PI3K/AKT signal pathway.
Journal of cellular biochemistry (2018-12-26)
Xiaohong Yin, Zhiyong Liu, Pan Zhu, Yuelong Wang, Qingqing Ren, Hongxu Chen, Jianguo Xu
ABSTRACT
Adamantinomatous craniopharyngiomas (adaCP) accounts for 5.6% to 15% of intracranial tumors. High expression of chemokine (C-X-C motif) ligand 12 (CXCL12, also known as stromal cell-derived factor 1 [SDF1]) and its receptor CXC receptor type 4 (CXCR4) are widespread in various malignancy via multiple signal transduction pathways. This study aims to investigate the mechanism of CXCL12/CXCR4 promoting proliferation, migration, and invasion of adaCP. Quantitative real-time polymerase chain reaction, Western blot analysis, and immunohistochemistry were used to evaluate the expression of CXCL12/CXCR4 mRNA and protein in 10 human adaCP tissues. Three successfully primary cell lines were obtained from native mainly solid tumor specimens, and confirmed by the means of inverted contrast microscope directly and following hematoxylin and eosin staining. Immunofluorescence was used to detect protein expression in vivo for the verification of primary cell line. Proliferation, migration, and invasion assays were performed to assess the biological functional role of CXCL12/CXCR4 in adaCP. The signal pathways involved in the action of CXCL12/CXCR4 in adaCP were also evaluated. CXCL12 and CXCR4 were highly expressed in human adaCP samples. Primary adaCP cells were isolated and detected by the means of immunofluorescence for the detection of pan cytokeratin (pan-CK) and vimentin (VIM). Overexpression of CXCL12/CXCR4 significantly promoted the proliferation, migration, and invasion of primary adaCP cells. Moreover, cancer-promoting activity of CXCL12/CXCR4 is partially through its facilitation of PI3K/AKT signal pathway. Our data showed that CXCL12/CXCR4 promotes adaCP proliferation, migration, and invasion through PI3K/AKT signal pathway. These findings suggested that therapeutic strategies regulating CXCL12/CXCR4 expression may provide an effective treatment of adaCP.