Merck
  • Oxytocin inhibits methamphetamine-associated learning and memory alterations by regulating DNA methylation at the Synaptophysin promoter.
Oxytocin inhibits methamphetamine-associated learning and memory alterations by regulating DNA methylation at the Synaptophysin promoter.
Addiction biology (2018-12-27)
Xin-Yu Fan, Jing-Yu Yang, Ying-Xu Dong, Ying Hou, Shuai Liu, Chun-Fu Wu
ABSTRACT
Methamphetamine (METH) causes memory changes, but the underlying mechanisms are poorly understood. Epigenetic mechanisms, including DNA methylation, can potentially cause synaptic changes in the brain. Oxytocin (OT) plays a central role in learning and memory, but little is known of the impact of OT on METH-associated memory changes. Here, we explored the role of OT in METH-induced epigenetic alterations that underlie spatial and cognitive memory changes. METH (2.0 mg/kg, i.p.) was administered to male C57BL/6 mice once every other day for 8 days. OT (2.5 μg, i.c.v.) or aCSF was given prior to METH. Spatial and cognitive memory were assessed. In Hip and PFC, synaptic structures and proteins were examined, levels of DNA methyltransferases (DNMTs) and methyl CpG binding protein 2 (MECP2) were determined, and the DNA methylation status at the Synaptophysin (Syn) promoter was assessed. METH enhanced spatial memory, decreased synapse length, downregulated DNMT1, DNMT3A, DNMT3B, and MECP2, and induced DNA hypomethylation at the Syn promoter in Hip. In contrast, METH reduced cognitive memory, increased synapse thickness, upregulated DNMT1, DNMT3A, and MECP2, and induced DNA hypermethylation at the Syn promoter in PFC. OT pretreatment specifically ameliorated METH-induced learning and memory alterations, normalized synapse structures, and regulated DNMTs and MECP2 to reverse the DNA methylation status changes at the Syn promoter in Hip and PFC. DNA methylation is an important gene regulatory mechanism underlying METH-induced learning and memory alterations. OT can potentially be used to specifically manipulate METH-related memory changes.
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 6G6-1C9, clone 6G6-1C9, Chemicon®, from mouse