Merck
MINSTED nanoscopy enters the Ångström localization range.
Nature biotechnology (2022-11-08)
Michael Weber, Henrik von der Emde, Marcel Leutenegger, Philip Gunkel, Sivakumar Sambandan, Taukeer A Khan, Jan Keller-Findeisen, Volker C Cordes, Stefan W Hell
ABSTRACT
Super-resolution techniques have achieved localization precisions in the nanometer regime. Here we report all-optical, room temperature localization of fluorophores with precision in the Ångström range. We built on the concept of MINSTED nanoscopy where precision is increased by encircling the fluorophore with the low-intensity central region of a stimulated emission depletion (STED) donut beam while constantly increasing the absolute donut power. By blue-shifting the STED beam and separating fluorophores by on/off switching, individual fluorophores bound to a DNA strand are localized with σ = 4.7 Å, corresponding to a fraction of the fluorophore size, with only 2,000 detected photons. MINSTED fluorescence nanoscopy with single-digit nanometer resolution is exemplified by imaging nuclear pore complexes and the distribution of nuclear lamin in mammalian cells labeled by transient DNA hybridization. Because our experiments yield a localization precision σ = 2.3 Å, estimated for 10,000 detected photons, we anticipate that MINSTED will open up new areas of application in the study of macromolecular complexes in cells.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder
Sigma-Aldrich
Pyranose Oxidase from Coriolus sp., recombinant, expressed in E. coli, ≥2.7 units/mg solid
Sigma-Aldrich
Anti-Vimentin antibody, Mouse monoclonal, clone V9, purified from hybridoma cell culture
Roche
Streptavidin, recombinant, liquid, ≥17 units/mg protein, pH range 5.0-9.0