Merck
Regulation of mitochondrial proteostasis by the proton gradient.
The EMBO journal (2022-08-02)
Maria Patron, Daryna Tarasenko, Hendrik Nolte, Lara Kroczek, Mausumi Ghosh, Yohsuke Ohba, Yvonne Lasarzewski, Zeinab Alsadat Ahmadi, Alfredo Cabrera-Orefice, Akinori Eyiama, Tim Kellermann, Elena I Rugarli, Ulrich Brandt, Michael Meinecke, Thomas Langer
ABSTRACT
Mitochondria adapt to different energetic demands reshaping their proteome. Mitochondrial proteases are emerging as key regulators of these adaptive processes. Here, we use a multiproteomic approach to demonstrate the regulation of the m-AAA protease AFG3L2 by the mitochondrial proton gradient, coupling mitochondrial protein turnover to the energetic status of mitochondria. We identify TMBIM5 (previously also known as GHITM or MICS1) as a Ca2+ /H+ exchanger in the mitochondrial inner membrane, which binds to and inhibits the m-AAA protease. TMBIM5 ensures cell survival and respiration, allowing Ca2+ efflux from mitochondria and limiting mitochondrial hyperpolarization. Persistent hyperpolarization, however, triggers degradation of TMBIM5 and activation of the m-AAA protease. The m-AAA protease broadly remodels the mitochondrial proteome and mediates the proteolytic breakdown of respiratory complex I to confine ROS production and oxidative damage in hyperpolarized mitochondria. TMBIM5 thus integrates mitochondrial Ca2+ signaling and the energetic status of mitochondria with protein turnover rates to reshape the mitochondrial proteome and adjust the cellular metabolism.
Sigma-Aldrich
Emetine dihydrochloride