Merck
Imaging Blood Vessels and Lymphatics in Mouse Trachea Wholemounts.
Methods in molecular biology (Clifton, N.J.) (2022-02-01)
Peter Baluk, Donald M McDonald
ABSTRACT
Changes in blood vessels and lymphatics in health and disease are easier to understand and interpret when studied microscopically in three dimensions. The mouse trachea is a simple, yet powerful, and versatile model system in which to achieve this. We describe practical immunohistochemical methods for fluorescence and confocal microscopy of wholemounts of the mouse trachea to achieve this purpose in which the entire vasculature can be visualized from the organ level to the cellular and subcellular level. Blood vessels and lymphatics have highly stereotyped vascular architectures that repeat in arcades between the tracheal cartilages. Arterioles, capillaries, and venules can be easily identified for the blood vessels, while the lymphatics consist of initial lymphatics and collecting lymphatics. Even small abnormalities in either blood vessels or lymphatics can be noticed and evaluated in three dimensions. We and others have used the mouse trachea for examining in situ angiogenesis and lymphangiogenesis, vascular development and regression, vessel patency, differences in transgenic mice, and pathological changes, such as increased vascular permeability induced by inflammatory mediators.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-VCAM-1 (CD106) Antibody, clone 6C7.1, clone 6C7.1, from rat
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-Clara Cell Secretory Protein Antibody, serum, Upstate®