Merck
  • Airway Smooth Muscle Cell Mitochondria Damage and Mitophagy in COPD via ERK1/2 MAPK.
Airway Smooth Muscle Cell Mitochondria Damage and Mitophagy in COPD via ERK1/2 MAPK.
International journal of molecular sciences (2022-11-27)
Lei Fang, Ming Zhang, Junling Li, Liang Zhou, Michael Tamm, Michael Roth
ABSTRACT
Chronic obstructive pulmonary disease (COPD) is characterized by irreversible deterioration of the airway wall. Cigarette smoking is the major trigger, and in vitro studies showed that cigarette smoke extract (CSE) induced mitophagy in airway epithelial cells via oxidative stress, but this mechanism was not studied in airway smooth muscle cells (ASMCs). Primary ASMCs isolated from COPD patients or non-disease donors were investigated for CSE-induced remodeling and mitochondria structure. Proteins were assessed by Western blots for remodeling: collagen type-I, α-smooth muscle actin (α-SMA) and fibronectin; autophagy: beclin-1, protein62 (p62), light chain (LC)3A/B; mitochondria activity: mitochondrially encoded cytochrome c oxidase II & -IV (MTCO2, MTCO4), peroxisome proliferator activated receptor gamma coactivator 1α (PGC-1α); lysosomes: early endosome antigen 1, lysosome activated membrane protein 1; and cell signaling: extracellular signal regulated kinase (ERK1/2). Lysotracker and Mitotracker were used to monitor mitochondria morphology and organelle co-localization. Compared with controls, untreated COPD ASMCs showed lower collagen type-I and α-SMA expressions, but increased fibronectin levels. CSE further downregulated collagen type-I and α-SMA expression, but upregulated fibronectin. CSE decreased PGC-1α, MTCO2, and MTCO4, but increased beclin-1, p62, and LC3. CSE upregulated mitophagy and lysosomes activity via ERK1/2 phosphorylation. In vitro, cigarette smoke induced the deterioration of ASMCs, which might explain the tissue loss and structural remodeling in COPD bronchi. The results suggest that preventing exceeded mitophagy in ASMCs might present a novel therapeutic target for COPD.