Merck
  • Interactions Between Epidermal Keratinocytes, Dendritic Epidermal T-Cells, and Hair Follicle Stem Cells.
Interactions Between Epidermal Keratinocytes, Dendritic Epidermal T-Cells, and Hair Follicle Stem Cells.
Methods in molecular biology (Clifton, N.J.) (2018-06-14)
Krithika Badarinath, Abhik Dutta, Akshay Hegde, Neha Pincha, Rupali Gund, Colin Jamora
ABSTRACT
The interplay of immune cells and stem cells in maintaining skin homeostasis and repair is an exciting new frontier in cutaneous biology. With the growing appreciation of the importance of this new crosstalk comes the requirement of methods to interrogate the molecular underpinnings of these leukocyte-stem cell interactions. Here we describe how a combination of FACS, cellular coculture assays, and conditioned media treatments can be utilized to advance our understanding of this emerging area of intercellular communication between immune cells and stem cells.