Merck
  • Suppression of MYC by PI3K/AKT/mTOR pathway inhibition in combination with all-trans retinoic acid treatment for therapeutic gain in acute myeloid leukaemia.
Suppression of MYC by PI3K/AKT/mTOR pathway inhibition in combination with all-trans retinoic acid treatment for therapeutic gain in acute myeloid leukaemia.
British journal of haematology (2022-04-26)
Sven Stengel, Kevin R Petrie, Yordan Sbirkov, Clara Stanko, Faezeh Ghazvini Zadegan, Veronica Gil, Rafał Skopek, Paweł Kamiński, Łukasz Szymański, Annamaria Brioli, Arthur Zelent, Tino Schenk
ABSTRACT
Aberrant activity of the phosphatidylinositol-3 kinase/protein kinase B/mammalian target of rapamycin (PI3K/AKT/mTOR [PAM]) pathway, as well as suppressed retinoic acid signalling, contribute to enhanced proliferation and the differentiation blockade of immature myeloid cells in acute myeloid leukaemia (AML). Inhibition of the PAM pathway was shown to affect especially mixed-lineage leukaemia-rearranged AML. Here, we sought to test a combined strategy using small molecule inhibitors against members of the PAM signalling pathway in conjunction with all-trans retinoic acid (ATRA) to target a larger group of different AML subtypes. We find that ATRA treatment in combination with inhibition of PI3K (ZSTK474), mTOR (WYE132) or PI3K/mTOR (BEZ235, dactolisib) drastically reduces protein levels of the proto-oncogene MYC. In combination with BEZ235, ATRA treatment led to almost complete eradication of cellular MYC, G1 arrest, loss of clonal capacity and terminal granulocytic differentiation. We demonstrate that PAM inhibitor/ATRA treatment targets MYC via independent mechanisms. While inhibition of the PAM pathway causes MYC phosphorylation at threonine 58 via glycogen synthase kinase 3 beta and subsequent degradation, ATRA reduces its expression. Here, we present an approach using a combination of known drugs to synergistically reduce aberrant MYC levels, thereby effectively blocking proliferation and enabling differentiation in various AML subtypes.