Merck
  • Functional analyses of STIM1 mutations reveal a common pathomechanism for tubular aggregate myopathy and Stormorken syndrome.
Functional analyses of STIM1 mutations reveal a common pathomechanism for tubular aggregate myopathy and Stormorken syndrome.
Neuropathology : official journal of the Japanese Society of Neuropathology (2020-10-20)
Georges Arielle Peche, Coralie Spiegelhalter, Roberto Silva-Rojas, Jocelyn Laporte, Johann Böhm
ABSTRACT
Tubular aggregate myopathy (TAM) is a progressive disorder characterized by muscle weakness, cramps, and myalgia. TAM clinically overlaps with Stormorken syndrome (STRMK), combining TAM with miosis, thrombocytopenia, hyposplenism, ichthyosis, short stature, and dyslexia. TAM and STRMK arise from gain-of-function mutations in STIM1 (stromal interaction molecule 1) or ORAI1, both encoding key regulators of Ca2+ homeostasis, and mutations in either gene result in excessive extracellular Ca2+ entry. The pathomechanistic similarities and differences between TAM and STRMK are only partially understood. Here we provide functional in vitro experiments demonstrating that STIM1 harboring the TAM D84G or the STRMK R304W mutation similarly cluster and exert a dominant effect on the wild-type protein. Both mutants recruit ORAI1 to the clusters, increase cytosolic Ca2+ levels, promote major nuclear import of the Ca2+ -dependent transcription factor NFAT (nuclear factor of activated T cells), and trigger the formation of circular membrane stacks. In conclusion, the analyzed TAM and STRMK mutations have a comparable impact on STIM1 protein function and downstream effects of excessive Ca2+ entry, highlighting that TAM and STRMK involve a common pathomechanism.
Sigma-Aldrich
Anti-STIM1 Antibody, Chemicon®, from rabbit