Merck
  • Phosphoproteomic analysis of neoadjuvant breast cancer suggests that increased sensitivity to paclitaxel is driven by CDK4 and filamin A.
Phosphoproteomic analysis of neoadjuvant breast cancer suggests that increased sensitivity to paclitaxel is driven by CDK4 and filamin A.
Nature communications (2022-12-09)
S Mouron, M J Bueno, A Lluch, L Manso, I Calvo, J Cortes, J A Garcia-Saenz, M Gil-Gil, N Martinez-Janez, J V Apala, E Caleiras, Pilar Ximénez-Embún, J Muñoz, L Gonzalez-Cortijo, R Murillo, R Sánchez-Bayona, J M Cejalvo, G Gómez-López, C Fustero-Torre, S Sabroso-Lasa, N Malats, M Martinez, A Moreno, D Megias, M Malumbres, R Colomer, M Quintela-Fandino
ABSTRACT
Precision oncology research is challenging outside the contexts of oncogenic addiction and/or targeted therapies. We previously showed that phosphoproteomics is a powerful approach to reveal patient subsets of interest characterized by the activity of a few kinases where the underlying genomics is complex. Here, we conduct a phosphoproteomic screening of samples from HER2-negative female breast cancer receiving neoadjuvant paclitaxel (N = 130), aiming to find candidate biomarkers of paclitaxel sensitivity. Filtering 11 candidate biomarkers through 2 independent patient sets (N = 218) allowed the identification of a subgroup of patients characterized by high levels of CDK4 and filamin-A who had a 90% chance of achieving a pCR in response to paclitaxel. Mechanistically, CDK4 regulates filamin-A transcription, which in turn forms a complex with tubulin and CLIP-170, which elicits increased binding of paclitaxel to microtubules, microtubule acetylation and stabilization, and mitotic catastrophe. Thus, phosphoproteomics allows the identification of explainable factors for predicting response to paclitaxel.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Cdk4 Antibody, clone DCS-35, clone DCS-35, Chemicon®, from mouse