Merck
  • High expression of glutamate-ammonia ligase is associated with unfavorable prognosis in patients with ovarian cancer.
High expression of glutamate-ammonia ligase is associated with unfavorable prognosis in patients with ovarian cancer.
Journal of cellular biochemistry (2018-03-27)
Shaohua Fan, Yanyan Wang, Zifeng Zhang, Jun Lu, Zhiyong Wu, Qun Shan, Chunhui Sun, Dongmei Wu, Mengqiu Li, Ning Sheng, Ying Xie, Yuanlin Zheng
ABSTRACT
Glutamate-ammonia ligase (GLUL), which is also called GS (glutamine synthetase), is the enzyme that catalyzes the synthesis of glutamine from glutamate and ammonia in an ATP-dependent reaction. Here, we found higher expression of GLUL in the ovarian cancer patients was associated with worse disease-free survival (DFS) and overall survival (OS). In addition, GLUL was heterogeneously expressed in various ovarian cancer cells. The mRNA and protein expression levels of GLUL in NIH:OVCAR-3 and ES-2 cells were obviously higher than that in the other types of ovarian cancer cells. Knockdown of GLUL in NIH:OVCAR-3 or ES-2 cells could significantly decrease the proliferation ability. Furthermore, GLUL knockdown markedly inhibited the p38 MAPK signaling pathway in NIH:OVCAR-3 or ES-2 cells. Our findings suggest that decreasing expression of GLUL may be a new approach that can be used for ovarian cancer treatment.