Merck
  • Localization of amylin-like immunoreactivity in the striped velvet gecko pancreas.
Localization of amylin-like immunoreactivity in the striped velvet gecko pancreas.
Anatomia, histologia, embryologia (2018-01-10)
H Suzuki, T Yamamoto
ABSTRACT
Immunohistochemical techniques were employed to investigate the distribution of amylin-like immunoreactive cells in the pancreas of gecko Homopholis fasciata. Four types of endocrine cells were distinguished: insulin immunoreactive (B cells), pancreatic polypeptide immunoreactive (PP cells), glucagon and pancreatic polypeptide immunoreactive (A/PP cells) and somatostatin immunoreactive cells (D cells). Pancreatic islets contained B, A/PP and D cells, whereas extrainsular regions contained B, D and PP cells. In the pancreatic islets, amylin-like immunoreactive cells corresponded to B cells, but not to A/PP or D cells. In the extrainsular regions, amylin-like immunoreactive cells corresponded to either B or PP cells. Amylin secreted from intrainsular B cells may regulate pancreatic hormone secretion in an autocrine and/or a paracrine fashion. On the other hand, amylin secreted from extrainsular PP and B cells, and/or intrainsular B cells may participate in the modulation of calcium homoeostasis in an endocrine fashion.
Sigma-Aldrich
Donkey Anti-Goat IgG Antibody, Rhodamine conjugate, Species Adsorbed, Chemicon®, from donkey