Merck
  • Identification of Novel Autophagy Inhibitors via Cell-Based High-Content Screening.
Identification of Novel Autophagy Inhibitors via Cell-Based High-Content Screening.
Methods in molecular biology (Clifton, N.J.) (2018-02-23)
Georgios Konstantinidis, Sonja Sievers, Yao-Wen Wu
ABSTRACT
Autophagy is a fundamental cellular catabolic pathway mediating the recycling of cellular components. Autophagy has been implicated in pathogenesis of diverse diseases such as neurodegeneration and cancer. Due to the therapeutic potential, the autophagy-modulating agents have profoundly enriched the spectrum of tools used to investigate autophagy. However, many of these compounds have additional off-target effects that may confound elucidation of autophagy in certain contexts. There remains high demand for highly specific and novel chemotypes that can be used to study the regulation mechanism of autophagy and contribute novel pharmacophores for therapeutic purposes. Here, we describe a cell-based quantitative high-content screening (HCS) for autophagy inhibitors using a human breast adenocarcinoma MCF7 cell line stably expressing EGFP-LC3, a bona fide marker of autophagy.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red