Merck
  • Deep cervical lymph node ligation aggravates AD-like pathology of APP/PS1 mice.
Deep cervical lymph node ligation aggravates AD-like pathology of APP/PS1 mice.
Brain pathology (Zurich, Switzerland) (2018-09-08)
Linmei Wang, Yanli Zhang, Ying Zhao, Charles Marshall, Ting Wu, Ming Xiao
ABSTRACT
The imbalance between production and clearance of amyloid-beta (Aβ) is a key step in the onset and development of Alzheimer's disease (AD). Therefore, reducing Aβ accumulation in the brain is a promising therapeutic strategy for AD. The recently discovered glymphatic system and meningeal lymphatic vasculature have been shown to be critical for the elimination of interstitial waste products, especially Aβ, from the brain. In the present study, ligation of deep cervical lymph nodes was performed to block drainage of this system and explore the consequences on Aβ-related pathophysiology. Five-month-old APP/PS1 mice and their wild-type littermates received deep cervical lymphatic node ligation. One month later, behavioral testing and pathological analysis were conducted. Results demonstrated that ligation of dcLNs exacerbated AD-like phenotypes of APP/PS1 mice, showing more severe brain Aβ accumulation, neuroinflammation, synaptic protein loss, impaired polarization of aquaporin-4 and deficits in cognitive and exploratory behaviors. These results suggest that brain lymphatic clearance malfunction is one of the deteriorating factors in the progression of AD, and restoring its function is a potential therapeutic target against AD.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-BACE Antibody, CT, clone 61-3E7, clone 61-3E7, Chemicon®, from mouse
Sigma-Aldrich
Anti-ADAM 10 Antibody, CT, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Interleukin-1β Antibody, Chemicon®, from rabbit