Merck
  • Arhgef6 (alpha-PIX) cytoskeletal regulator signals to GTPases and Cofilin to couple T cell migration speed and persistence.
Arhgef6 (alpha-PIX) cytoskeletal regulator signals to GTPases and Cofilin to couple T cell migration speed and persistence.
Journal of leukocyte biology (2021-02-03)
Dejan Mamula, Mark Korthals, Johannes Hradsky, Anna Gottfried, Klaus-Dieter Fischer, Kerry Tedford
ABSTRACT
Immunity is governed by successful T cell migration, optimized to enable a T cell to fully scan its environment without wasted movement by balancing speed and turning. Here we report that the Arhgef6 RhoGEF (aka alpha-PIX; αPIX; Cool-2), an activator of small GTPases, is required to restrain cell migration speed and cell turning during spontaneous migration on 2D surfaces. In Arhgef6-/- T cells, expression of Arhgef7 (beta-PIX; βPIX; Cool-1), a homolog of Arhgef6, was increased and correlated with defective activation and localization of Rac1 and CDC42 GTPases, respectively. Downstream of Arhgef6, PAK2 (p21-activated kinase 2) and LIMK1 phosphorylation was reduced, leading to increased activation of Cofilin, the actin-severing factor. Consistent with defects in these signaling pathways, Arhgef6-/- T cells displayed abnormal bilobed lamellipodia and migrated faster, turned more, and arrested less than wild-type (WT) T cells. Using pharmacologic inhibition of LIMK1 (LIM domain kinase 1) to induce Cofilin activation in WT T cells, we observed increased migration speed but not increased cell turning. In contrast, inhibition of Cdc42 increased cell turning but not speed. These results suggested that the increased speed of the Arhgef6-/- T cells is due to hyperactive Cofilin while the increased turning may be due to abnormal GTPase activation and recruitment. Together, these findings reveal that Arhgef6 acts as a repressor of T cell speed and turning by limiting actin polymerization and lamellipodia formation.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse