Merck
  • Duloxetine attenuates pain in association with downregulation of platelet serotonin transporter in patients with burning mouth syndrome and atypical odontalgia.
Duloxetine attenuates pain in association with downregulation of platelet serotonin transporter in patients with burning mouth syndrome and atypical odontalgia.
Human psychopharmacology (2021-09-21)
Mariko Nakamura, Akira Yoshimi, Akihiro Mouri, Tatsuya Tokura, Hiroyuki Kimura, Shinichi Kishi, Tomoya Miyauchi, Kunihiro Iwamoto, Mikiko Ito, Aiji Sato-Boku, Norio Ozaki, Toshitaka Nabeshima, Yukihiro Noda
ABSTRACT
The aim of this study was evaluation of the association between severity of pain and expression of total or ubiquitinated serotonin transporter (SERT) protein in patients with burning mouth syndrome and atypical odontalgia (BMS/AO), who were treated by duloxetine. Patients with BMS/AO were assessed for severity of pain using the visual analog scale (VAS), and expression of total and ubiquitinated SERT protein in platelets before (baseline) and 12 weeks after duloxetine-treatment. The expression of total and ubiquitinated SERT protein at baseline in all patients (n = 33) were higher and lower, respectively, compared to those in healthy controls. 12 weeks after duloxetine-treatment, there was no difference in the total SERT protein levels between patients (n = 21) and healthy controls. In the 16 patients who could be measured, mean VAS scores and total SERT protein levels were significantly decreased after the treatment, compared to those at baseline. There was tendency for a positive correlation between total SERT protein levels and VAS scores in these patients. Our findings indicate that duloxetine relieves pain in association with downregulation of platelet SERT expression in patients with BMS/AO.