Merck
  • A conserved enzyme of smut fungi facilitates cell-to-cell extension in the plant bundle sheath.
A conserved enzyme of smut fungi facilitates cell-to-cell extension in the plant bundle sheath.
Nature communications (2022-10-13)
Bilal Ökmen, Elaine Jaeger, Lena Schilling, Natalie Finke, Amy Klemd, Yoon Joo Lee, Raphael Wemhöner, Markus Pauly, Ulla Neumann, Gunther Doehlemann
ABSTRACT
Smut fungi comprise one of the largest groups of fungal plant pathogens causing disease in all cereal crops. They directly penetrate host tissues and establish a biotrophic interaction. To do so, smut fungi secrete a wide range of effector proteins, which suppress plant immunity and modulate cellular functions as well as development of the host, thereby determining the pathogen's lifestyle and virulence potential. The conserved effector Erc1 (enzyme required for cell-to-cell extension) contributes to virulence of the corn smut Ustilago maydis in maize leaves but not on the tassel. Erc1 binds to host cell wall components and displays 1,3-β-glucanase activity, which is required to attenuate β-glucan-induced defense responses. Here we show that Erc1 has a cell type-specific virulence function, being necessary for fungal cell-to-cell extension in the plant bundle sheath and this function is fully conserved in the Erc1 orthologue of the barley pathogen Ustilago hordei.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Gelatin from cold water fish skin, 40-50% in H2O
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid