Merck
  • De novo sphingolipid biosynthesis necessitates detoxification in cancer cells.
De novo sphingolipid biosynthesis necessitates detoxification in cancer cells.
Cell reports (2022-09-29)
Meghan E Spears, Namgyu Lee, Sunyoung Hwang, Sung Jin Park, Anne E Carlisle, Rui Li, Mihir B Doshi, Aaron M Armando, Jenny Gao, Karl Simin, Lihua Julie Zhu, Paul L Greer, Oswald Quehenberger, Eduardo M Torres, Dohoon Kim
ABSTRACT
Sphingolipids play important signaling and structural roles in cells. Here, we find that during de novo sphingolipid biosynthesis, a toxic metabolite is formed with critical implications for cancer cell survival. The enzyme catalyzing the first step in this pathway, serine palmitoyltransferase complex (SPT), is upregulated in breast and other cancers. SPT is dispensable for cancer cell proliferation, as sphingolipids can be salvaged from the environment. However, SPT activity introduces a liability as its product, 3-ketodihydrosphingosine (3KDS), is toxic and requires clearance via the downstream enzyme 3-ketodihydrosphingosine reductase (KDSR). In cancer cells, but not normal cells, targeting KDSR induces toxic 3KDS accumulation leading to endoplasmic reticulum (ER) dysfunction and loss of proteostasis. Furthermore, the antitumor effect of KDSR disruption can be enhanced by increasing metabolic input (via high-fat diet) to allow greater 3KDS production. Thus, de novo sphingolipid biosynthesis entails a detoxification requirement in cancer cells that can be therapeutically exploited.
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma