Merck
  • Segregation of the membrane cargoes, BACE1 and amyloid precursor protein (APP) throughout the Golgi apparatus.
Segregation of the membrane cargoes, BACE1 and amyloid precursor protein (APP) throughout the Golgi apparatus.
Traffic (Copenhagen, Denmark) (2022-01-26)
Lou Fourriere, Ellie Hyun-Jung Cho, Paul A Gleeson
ABSTRACT
The intracellular trafficking of β-site amyloid precursor protein (APP) cleaving enzyme (BACE1) and APP regulates amyloid-β production. Our previous work demonstrated that newly synthesized BACE1 and APP are segregated into distinct trafficking pathways from the trans-Golgi network (TGN), and that alterations in their trafficking lead to an increase in Aβ production in non-neuronal and neuronal cells. However, it is not known whether BACE1 and APP are transported through the Golgi stacks together and sorted at the TGN or segregated prior to arrival at the TGN. To address this question, we have used high-resolution Airyscan technology followed by Huygens deconvolution to quantify the overlap of BACE1 and APP in Golgi subcompartments in HeLa cells and primary neurons. Here, we show that APP and BACE1 are segregated, on exit from the endoplasmic reticulum and in the cis-Golgi and throughout the Golgi stack. In contrast, the transferrin receptor, which exits the TGN in AP-1 mediated transport carriers as for BACE1, colocalizes with BACE1, but not APP, throughout the Golgi stack. The segregation of APP and BACE1 is independent of the Golgi ribbon structure and the cytoplasmic domain of the cargo. Overall, our findings reveal the segregation of different membrane cargoes early in the secretory pathway, a finding relevant to the regulation of APP processing events.
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Amyloid β Antibody, clone W0-2, clone WO2, from mouse
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal (751-770) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
DAPI, for nucleic acid staining