Merck
  • INF2-mediated actin filament reorganization confers intrinsic resilience to neuronal ischemic injury.
INF2-mediated actin filament reorganization confers intrinsic resilience to neuronal ischemic injury.
Nature communications (2022-10-14)
Barbara Calabrese, Steven L Jones, Yoko Shiraishi-Yamaguchi, Michael Lingelbach, Uri Manor, Tatyana M Svitkina, Henry N Higgs, Andy Y Shih, Shelley Halpain
ABSTRACT
During early ischemic brain injury, glutamate receptor hyperactivation mediates neuronal death via osmotic cell swelling. Here we show that ischemia and excess NMDA receptor activation cause actin to rapidly and extensively reorganize within the somatodendritic compartment. Normally, F-actin is concentrated within dendritic spines. However, <5 min after bath-applied NMDA, F-actin depolymerizes within spines and polymerizes into stable filaments within the dendrite shaft and soma. A similar actinification occurs after experimental ischemia in culture, and photothrombotic stroke in mouse. Following transient NMDA incubation, actinification spontaneously reverses. Na+, Cl-, water, and Ca2+ influx, and spine F-actin depolymerization are all necessary, but not individually sufficient, for actinification, but combined they induce activation of the F-actin polymerization factor inverted formin-2 (INF2). Silencing of INF2 renders neurons vulnerable to cell death and INF2 overexpression is protective. Ischemia-induced dendritic actin reorganization is therefore an intrinsic pro-survival response that protects neurons from death induced by cell edema.
Sigma-Aldrich
(−)-Blebbistatin, solid, synthetic
Sigma-Aldrich
C646, ≥98% (HPLC)
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Nckβ Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-INF2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-NeuN Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Bumetanide, ≥98%