Merck
Protein tyrosine kinase 6 regulates activation of SRC kinase.
The Journal of biological chemistry (2022-10-14)
Wanian M Alwanian, Katarina Vlajic, Wenjun Bie, Andre Kajdacsy-Balla, Angela L Tyner
ABSTRACT
Expression of Protein tyrosine kinase 6 (PTK6) is upregulated in several human solid tumors, and it has oncogenic roles in prostate and breast cancer. PTK6 and SRC kinase are distantly related, share many substrates, and often regulate the same signaling pathways, but whether they interact to regulate signaling is not well understood. We characterized crosstalk between PTK6 and SRC and show that PTK6 can directly phosphorylate SRC to promote its activation. Stable knockdown of PTK6 in multiple cancer cell lines leads to decreased activating phosphorylation of SRC. We show that coexpression of kinase-dead SRC and active PTK6 in mouse embryonic fibroblasts lacking Src, Yes, and Fyn results in activating phosphorylation of SRC. However, there is no reciprocal effect, because active SRC does not promote activating phosphorylation of PTK6. Overexpression of active PTK6 maintained activation of epidermal growth factor receptor (EGFR), AKT, and FAK, but not SHP2 and ERK1/2 in cells with knockdown of SRC. Both PTK6 and SRC are regulated by EGFR, and its inhibition with erlotinib downregulated PTK6 and to a lesser degree SRC activation in LNCaP cells that overexpress active PTK6. Erlotinib treatment also led to AKT inhibition, but overexpression of active PTK6 prevented this. Our data demonstrate overlapping and unique functions for PTK6 and SRC. Finally, we show that PTK6 and SRC are coexpressed in subsets of human prostate and breast cancer cells, and active PTK6 and active SRC colocalize in prostate cancer, supporting a role for PTK6 in promoting SRC activity in cancer.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Epidermal Growth Factor from mouse, EGF
Sigma-Aldrich
Anti-phospho-Brk (Tyr342) Antibody, Upstate®, from rabbit