Merck
  • Receptor tyrosine kinases regulate signal transduction through a liquid-liquid phase separated state.
Receptor tyrosine kinases regulate signal transduction through a liquid-liquid phase separated state.
Molecular cell (2022-03-02)
Chi-Chuan Lin, Kin Man Suen, Polly-Anne Jeffrey, Lukasz Wieteska, Jessica A Lidster, Peng Bao, Alistair P Curd, Amy Stainthorp, Caroline Seiler, Hans Koss, Eric Miska, Zamal Ahmed, Stephen D Evans, Carmen Molina-París, John E Ladbury
ABSTRACT
The recruitment of signaling proteins into activated receptor tyrosine kinases (RTKs) to produce rapid, high-fidelity downstream response is exposed to the ambiguity of random diffusion to the target site. Liquid-liquid phase separation (LLPS) overcomes this by providing elevated, localized concentrations of the required proteins while impeding competitor ligands. Here, we show a subset of phosphorylation-dependent RTK-mediated LLPS states. We then investigate the formation of phase-separated droplets comprising a ternary complex including the RTK, (FGFR2); the phosphatase, SHP2; and the phospholipase, PLCγ1, which assembles in response to receptor phosphorylation. SHP2 and activated PLCγ1 interact through their tandem SH2 domains via a previously undescribed interface. The complex of FGFR2 and SHP2 combines kinase and phosphatase activities to control the phosphorylation state of the assembly while providing a scaffold for active PLCγ1 to facilitate access to its plasma membrane substrate. Thus, LLPS modulates RTK signaling, with potential consequences for therapeutic intervention.
Product Description
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.