Merck
  • Rational design of a sensitivity-enhanced tracer for discovering efficient APC-Asef inhibitors.
Rational design of a sensitivity-enhanced tracer for discovering efficient APC-Asef inhibitors.
Nature communications (2022-08-25)
Jie Zhong, Yuegui Guo, Shaoyong Lu, Kun Song, Ying Wang, Li Feng, Zhen Zheng, Qiufen Zhang, Jiacheng Wei, Peng Sang, Yan Shi, Jianfeng Cai, Guoqiang Chen, Chen-Ying Liu, Xiuyan Yang, Jian Zhang
ABSTRACT
The adenomatous polyposis coli (APC)-Rho guanine nucleotide exchange factor 4 (Asef) protein-protein interaction (PPI) is essential for colorectal cancer metastasis, making it a promising drug target. Herein, we obtain a sensitivity-enhanced tracer (tracer 7) with a high binding affinity (Kd = 0.078 μM) and wide signal dynamic range (span = 251 mp). By using tracer 7 in fluorescence-polarization assays for APC-Asef inhibitor screening, we discover a best-in-class inhibitor, MAI-516, with an IC50 of 0.041 ± 0.004 μM and a conjugated transcriptional transactivating sequence for generating cell-permeable MAIT-516. MAIT-516 inhibits CRC cell migration by specifically hindering the APC-Asef PPI. Furthermore, MAIT-516 exhibits no cytotoxic effects on normal intestinal epithelial cell and colorectal cancer cell growth. Overall, we develop a sensitivity-enhanced tracer for fluorescence polarization assays, which is used for the precise quantification of high-activity APC-Asef inhibitors, thereby providing insight into PPI drug development.
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-APC Antibody, NT, clone Ali 12.28, clone Ali 12.28, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)