Merck
  • Prohormone convertases 1/3 and 2 together orchestrate the site-specific cleavages of progastrin to release gastrin-34 and gastrin-17.
Prohormone convertases 1/3 and 2 together orchestrate the site-specific cleavages of progastrin to release gastrin-34 and gastrin-17.
The Biochemical journal (2008-06-17)
Jens F Rehfeld, Xiaorong Zhu, Christina Norrbom, Jens R Bundgaard, Anders H Johnsen, John E Nielsen, Jonas Vikesaa, Jeffrey Stein, Arunangsu Dey, Donald F Steiner, Lennart Friis-Hansen
ABSTRACT
Cellular synthesis of peptide hormones requires PCs (prohormone convertases) for the endoproteolysis of prohormones. Antral G-cells synthesize the most gastrin and express PC1/3, 2 and 5/6 in the rat and human. But the cleavage sites in progastrin for each PC have not been determined. Therefore, in the present study, we measured the concentrations of progastrin, processing intermediates and alpha-amidated gastrins in antral extracts from PC1/3-null mice and compared the results with those in mice lacking PC2 and wild-type controls. The expression of PCs was examined by immunocytochemistry and in situ hybridization of mouse G-cells. Finally, the in vitro effect of recombinant PC5/6 on progastrin and progastrin fragments containing the relevant dibasic cleavage sites was also examined. The results showed that mouse G-cells express PC1/3, 2 and 5/6. The concentration of progastrin in PC1/3-null mice was elevated 3-fold. Chromatography showed that cleavage of the Arg(36)Arg(37) and Arg(73)Arg(74) sites were grossly decreased. Accordingly, the concentrations of progastrin products were markedly reduced, alpha-amidated gastrins (-34 and -17) being 25% of normal. Lack of PC1/3 was without effect on the third dibasic site (Lys(53)Lys(54)), which is the only processing site for PC2. Recombinant PC5/6 did not cleave any of the dibasic processing sites in progastrin and fragments containing the relevant dibasic processing sites. The complementary cleavages of PC1/3 and 2, however, suffice to explain most of the normal endoproteolysis of progastrin. Moreover, the results show that PCs react differently to the same dibasic sequences, suggesting that additional structural factors modulate the substrate specificity.
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt, ≥99% (HPLC)