Merck
  • Reorganization of the Brain Extracellular Matrix in Hippocampal Sclerosis.
Reorganization of the Brain Extracellular Matrix in Hippocampal Sclerosis.
International journal of molecular sciences (2022-07-29)
Barbara Sitaš, Mihaela Bobić-Rasonja, Goran Mrak, Sara Trnski, Magdalena Krbot Skorić, Darko Orešković, Vinka Knezović, Željka Petelin Gadže, Zdravko Petanjek, Goran Šimić, Danijela Kolenc, Nataša Jovanov Milošević
ABSTRACT
The extracellular matrix (ECM) is an important regulator of excitability and synaptic plasticity, especially in its highly condensed form, the perineuronal nets (PNN). In patients with drug-resistant mesial temporal lobe epilepsy (MTLE), hippocampal sclerosis type 1 (HS1) is the most common histopathological finding. This study aimed to evaluate the ECM profile of HS1 in surgically treated drug-resistant patients with MTLE in correlation to clinical findings. Hippocampal sections were immunohistochemically stained for aggrecan, neurocan, versican, chondroitin-sulfate (CS56), fibronectin, Wisteria floribunda agglutinin (WFA), a nuclear neuronal marker (NeuN), parvalbumin (PV), and glial-fibrillary-acidic-protein (GFAP). In HS1, besides the reduced number of neurons and astrogliosis, we found a significantly changed expression pattern of versican, neurocan, aggrecan, WFA-specific glycosylation, and a reduced number of PNNs. Patients with a lower number of epileptic episodes had a less intense diffuse WFA staining in Cornu Ammonis (CA) fields. Our findings suggest that PNN reduction, changed ECM protein, and glycosylation expression pattern in HS1 might be involved in the pathogenesis and persistence of drug-resistant MTLE by contributing to the increase of CA pyramidal neurons' excitability. This research corroborates the validity of ECM molecules and their modulators as a potential target for the development of new therapeutic approaches to drug-resistant epilepsy.
Sigma-Aldrich
Lectin from Wisteria floribunda, buffered aqueous solution, biotin conjugate
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution