Merck
  • TDP1-independent pathways in the process and repair of TOP1-induced DNA damage.
TDP1-independent pathways in the process and repair of TOP1-induced DNA damage.
Nature communications (2022-07-23)
Huimin Zhang, Yun Xiong, Dan Su, Chao Wang, Mrinal Srivastava, Mengfan Tang, Xu Feng, Min Huang, Zhen Chen, Junjie Chen
ABSTRACT
Anticancer drugs, such as camptothecin (CPT), trap topoisomerase I (TOP1) on DNA and form TOP1 cleavage complexes (TOP1cc). Alternative repair pathways have been suggested in the repair of TOP1cc. However, how these pathways work with TDP1, a key repair enzyme that specifically hydrolyze the covalent bond between TOP1 catalytic tyrosine and the 3'-end of DNA and contribute to the repair of TOP1cc is poorly understood. Here, using unbiased whole-genome CRISPR screens and generation of co-deficient cells with TDP1 and other genes, we demonstrate that MUS81 is an important factor that mediates the generation of excess double-strand breaks (DSBs) in TDP1 KO cells. APEX1/2 are synthetic lethal with TDP1. However, deficiency of APEX1/2 does not reduce DSB formation in TDP1 KO cells. Together, our data suggest that TOP1cc can be either resolved directly by TDP1 or be converted into DSBs and repaired further by the Homologous Recombination (HR) pathway.
Sigma-Aldrich
Anti-Topoisomerase I-DNA Covalent Complexes Antibody, clone 1.1A, clone 1.1A, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
APE1 Inhibitor III, The APE1 Inhibitor III controls the biological activity of APE1. This small molecule/inhibitor is primarily used for Cell Structure applications.