Merck
  • lncRNA RP11-10A14.5: a potential prognosis biomarker for LUAD through regulation on proliferation and metastasis.
lncRNA RP11-10A14.5: a potential prognosis biomarker for LUAD through regulation on proliferation and metastasis.
Discover. Oncology (2022-05-17)
Zhijie Lin, Fenglan Feng, Jiaming Liang, Haikang Zeng, Jin Li
ABSTRACT
Lung cancer is the malignancy most commonly seen worldwide. Emerging evidences indicated that lncRNAs may serve as a prognosis marker and play important role in NSCLC tumor biology. In this work, we analyzed the prognosis value of RP11-10A14.5 using TCGA and GEPIA database and expression profiles using PCR and FISH assay. The biological roles of RP11-10A14.5 in cell growth and invasion were determined by in vitro and in vivo experiments. Expression of RP11-10A14.5 is correlated with increased clinical stage and poor survival prognosis. In vitro experiments revealed that RP11-10A14.5 was widely expressed in lung cancer cell lines and mainly distributed in the cytoplasm and enhanced the growth, invasion and migration ability of NSCLC cell lines. Immunofluorescence assay suggested that RP11-10A14.5 may promote EMT by downregulating E-cadherin and upregulating N-cadherin and Vimentin. Flow cytometry results suggested that RP11-10A14.5 did not significantly affect cell cycle function, but could significantly inhibit apoptosis which may further enhance metastasis cell survival. In conclusion, RP11-10A14.5 is associated with clinical stage and poor survival outcome, may serve as a diagnosis and prognosis predictor for LUAD. Further, RP11-10A14.5 could promote LUAD cell growth and metastasis.