Merck
Ion channels of nociception.
Annual review of physiology (1999-04-01)
E W McCleskey, M S Gold
ABSTRACT
Nociceptors are the first cells in the series of neurons that lead to the sensation of pain. The essential functions of nociceptors--transducing noxious stimuli into depolarizations that trigger action potentials, conducting the action potentials from the peripheral sensory site to the synapse in the central nervous system, and converting the action potentials into neurotransmitter release at the presynaptic terminal--all depend on ion channels. This review discusses recent results in the converging fields of nociception and ion channel biology. It focuses on (a) the capsaicin receptor and its possible role in thermosensation, (b) ATP-gated channels, (c) proton-gated channels, and (d) nociceptor-specific Na+ channels.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 70 purifications
Roche
Dispase® II (neutral protease, grade II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture